missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ TR-FRET LXR beta Coactivator Assay Kit, goat
Description
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15129:
The LanthaScreen™ TR-FRET LXR (liver X receptor) beta Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential LXR beta ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and an LXR beta ligand-binding domain (LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.
How it works:
In the LanthaScreen™ TR-FRET LXR beta Coactivator Assay, LXR beta-LBD is added to ligand test compounds followed by the addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the LXR beta-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET LXR beta Coactivator Assay Kit contains LXR beta-LBD (GST) protein, fluorescently labeled D22 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
The LanthaScreen™ TR-FRET LXR (liver X receptor) beta Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential LXR beta ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and an LXR beta ligand-binding domain (LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.
How it works:
In the LanthaScreen™ TR-FRET LXR beta Coactivator Assay, LXR beta-LBD is added to ligand test compounds followed by the addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the LXR beta-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET LXR beta Coactivator Assay Kit contains LXR beta-LBD (GST) protein, fluorescently labeled D22 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Specifications
Specifications
| Conjugate | Tb (Terbium) |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 800 x 20 μL assays |
| Packaging | 384-well Plate |
| Content And Storage | Store in ultra-cold freezer (-68 to -85°C). |
| Assay Entry | Biochemical coregulator interaction |
| Gene ID (Entrez) | 7376 |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?