missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET PPAR alpha Competitive Binding Assay Kit, rabbit

Product Code. 15343806
Change view
Click to view available options
Quantity:
400 x 40 μL assays, 400 x 20 μL Assays
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15343806 400 x 40 μL assays, 400 x 20 μL Assays 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15343806 Supplier Thermo Scientific™ Supplier No. A15143
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The LanthaScreen™ TR-FRET PPAR alpha Competitive Binding Assay provides a sensitive and robust method for high-throughput screening (HTS) of ligands for peroxisome proliferator-activated receptor alpha (PPAR alpha).

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4892:

The LanthaScreen™ TR-FRET PPAR alpha Competitive Binding Assay provides a sensitive and robust method for high-throughput screening (HTS) of ligands for peroxisome proliferator-activated receptor alpha (PPAR alpha). The kit uses a terbium-labeled anti-GST antibody, a fluorescent small-molecule pan-PPAR ligand (Fluormone™ Pan-PPAR Green), and a human PPAR alpha ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST), in a homogeneous mix-and-read assay format.

To assay:

When running the LanthaScreen™ TR-FRET PPAR alpha Competitive Binding Assay, Fluormone™ Pan-PPAR Green is added to ligand test compounds followed by addition of a mixture of the PPAR alpha-LBD and terbium anti-GST antibody. When the Fluormone™ Pan-PPAR Green is bound to PPAR alpha, energy transfer from the terbium-labeled antibody to the tracer occurs, and a high TR-FRET ratio is observed. Competitive ligand binding to PPAR alpha is detected by a test compound's ability to displace the tracer, which results in a loss of FRET between the antibody and the tracer. After an incubation period at room temperature, the 520/495 nm TR-FRET ratio is calculated and can be used to determine the IC50 from a dose response curve of the compound (Figure 1). This type of binding assay is analogous to radioligandbased assays, except that it eliminates handling of radioactivity and enables a homogeneous 'addition-only' format.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Conjugate Tb (Terbium)
Detection Method Fluorescence, Fluorescent
Description Color: Green
For Use With (Equipment) Microplate Reader
For Use With (Application) Nuclear Receptors, Nuclear Receptor Assay, TR-FRET
Quantity 400 x 40 μL assays, 400 x 20 μL Assays
Packaging 384-well Plate
Sample Type 384-well Plate
Content And Storage • Tb-anti-GST Antibody (Rabbit) (store at -20°C)
• PPAR alpha-LBD, GST (-80°C)
• Fluormone Pan-PPAR Green (-20°C)
• TR-FRET PPAR Assay Buffer (4°C)
• 1M DTT (-20°C or -80°C)
Assay Category Biochemical Assay
Druggable Target Nuclear Receptors
Readout TR-FRET (Binding)
Product Line LanthaScreen
Ligand Pan-PPAR
Product Type TR-FRET PPAR Alpha Competitive Binding Assay Kit
No. of Assays 400 x 40 μL Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.