missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ TR-FRET PPAR delta Competitive Binding Assay Kit, goat

Product Code. 10562894
Click to view available options
Quantity:
400 x 40 μL
Unit Size:
Pack of 400
This item is not returnable. View return policy

Product Code. 10562894

Brand: Invitrogen™ PV4893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ TR-FRET PPAR delta Competitive Binding Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15144:

The LanthaScreen™ TR-FRET PPAR delta Competitive Binding Assay provides a sensitive and robust method for high-throughput screening (HTS) of ligands for peroxisome proliferator-activated receptor delta (PPAR delta). The kit uses a terbium-labeled anti-GST antibody, a fluorescent small-molecule pan-PPAR ligand (Fluormone™ Pan-PPAR Green), and a human PPAR delta ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST), in a homogeneous mix-and-read assay format.

To assay:
When running the LanthaScreen™ TR-FRET PPAR delta Competitive Binding Assay, Fluormone™ Pan-PPAR Green is added to ligand test compounds, followed by addition of a mixture of the PPAR delta-LBD and terbium anti-GST antibody. When the Fluormone™ Pan-PPAR Green is bound to PPAR delta, energy transfer from the terbium-labeled antibody to the tracer occurs, and a high TR-FRET ratio is observed. Competitive ligand binding to PPAR delta is detected by a test compound’s ability to displace the tracer, which results in a loss of FRET between the antibody and the tracer. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the IC50 from a dose response curve of the compound (Figure 1). This type of binding assay is analogous to radioligandbased assays, except that it eliminates handling of radioactivity and enables a homogeneous, addition-only format.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 400 x 40 μL
Packaging 384-well Plate
Content And Storage The LanthaScreen™ TR-FRET PPAR delta Competitive Binding Assay Kit contains PPAR delta-LBD (GST) protein, Fluormone™ Pan-PPAR Green, terbium-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Assay Entry Biochemical competitive binding
Readout End Point
Shipping Condition Dry Ice
Target Entry PPAR delta, NR1C2, PPARD
Product Line LanthaScreen
For Use With (Application) Competitive Binding Assay, TR-FRET
Ligand Pan-PPAR
Product Type TR-FRET PPAR Delta Competitive Binding Assay Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.