missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ TR-FRET PPAR gamma Coactivator Assay Kit, rabbit

Product Code. 15329736
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15329736 800 x 20 μL assays 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15329736 Supplier Invitrogen™ Supplier No. A15126
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LanthaScreen™ TR-FRET PPAR gamma Coactivator Assay Kit, rabbit

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4548:

The LanthaScreen™ TR-FRET PPAR gamma Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential PPAR gamma ligands as agonists or antagonists of ligand-dependent coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a Peroxisome Proliferator Activated Receptor (PPAR) gamma ligand-binding domain (PPAR gamma-LBD) that is tagged with glutathione-S-transferase (GST), in a homogenous mix-and-read assay format.

Agonist mode
To run the LanthaScreen™ TR-FRET Peroxisome Proliferator Activated gamma Receptor Coactivator Assay in agonist mode (to identify agonist compounds), PPAR gamma-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After an incubation period at room temperature, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the PPAR gamma-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).

Antagonist mode
When the LanthaScreen™ TR-FRET Peroxisome Proliferator Activated gamma Receptor Coactivator Assay is run in antagonist mode (to identify antagonist compounds), PPAR gamma-LBD is added to ligand test compounds followed by addition of a mixture of agonist, fluores-cein-coactivator peptide, and Tb-anti-GST antibody. The concentration of agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode (Figure 2).

Contents and Storage:
The LanthaScreen™ TR-FRET PPARγ Coactivator Assay Kit contains PPAR gamma-LBD (GST) protein, fluorescently labeled TRAP220/ DRIP-2 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Conjugate Tb (Terbium)
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 800 x 20 μL assays
Packaging 384-well Plate
Content And Storage Tb-anti-GST Antibody (Rabbit): Store at -20 °C
PPAR gamma-LBD, GST: Store at -80 °C
Fluoroscein Coactivator Peptide: Store at -20 °C
TR-FRET Coregulatore Buffer F: Store at 4 °C
1M DTT: Store at -20 °C or -80 °C
Assay Category Biochemical Assay
Druggable Target Nuclear Receptors
Readout TR-FRET (Activity)
Product Line LanthaScreen
For Use With (Application) Nuclear Receptor Assay, TR-FRET
Ligand PPAR Gamma
Product Type TR-FRET PPAR Gamma Coactivator Assay Kit
No. of Assays 800 x 20 μL assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.