missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit, goat

Product Code. 10246793
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10246793 800 x 20 μL assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246793 Supplier Invitrogen™ Supplier No. PV4839
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit (Goat)

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15142:

The LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit provides a sensitive and robust method for high-throughput screening (HTS) of ligands for the pregnane X receptor (PXR), also known as the steroid and xenobiotic receptor (SXR). The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescent small-molecule PXR ligand (Fluormone™ PXR (SXR) Green), and a human PXR (SXR) ligand-binding domain that is tagged with glutathione S-transferase (GST) in a homogeneous mix-and-read assay format.

To assay:
When running the LanthaScreen™ TR-FRET PXR (SXR) competitive binding assay, a Tb-labeled anti-GST antibody is used to indirectly label the receptor by binding to its GST tag. Competitive ligand binding is detected by a test compound’s ability to displace a fluorescent ligand (tracer) from the receptor, which results in a loss of FRET signal between the Tb-anti-GST antibody and the tracer (Figure 1). This type of binding assay is analogous to radioligand-based assays, except that it eliminates handling of radioactivity and enables a homogeneous, addition-only format.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 800 x 20 μL assays
Packaging 384-well Plate
Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Assay Entry Biochemical competitive binding
Gene ID (Entrez) 8856
Readout End Point
Shipping Condition Dry Ice
Target Entry NR1I2, PXR(SXR)
Product Line LanthaScreen
For Use With (Application) Competitive Binding Assay, TR-FRET
Ligand PXR
Product Type TR-FRET PXR (SXR) Competitive Binding Assay Kit
No. of Assays 800 x 20 μL assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.