missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Lapatinib

Product Code. 18748056
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18748056 10 mg 10mg
18727506 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18748056 Supplier R&D Systems™ Supplier No. 6811/10
  • ONLINE EXCLUSIVE PRICE
    90.85 €
    81.90 € / 10mg
    Save 8.95 €
    9% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective dual ErbB2 (HER-2) and EGFR inhibitor (IC50 values are 9.8 and 10.2 nM, respectively)

Lapatinib is a potent and selective dual ErbB2 (HER-2) and EGFR inhibitor (IC50 values are 9.8 and 10.2 nM, respectively). Selective for EGFR and ErbB-2 versus other proliferative kinases. Inhibits proliferation in multiple cancer cell lines (average IC50 values <0.16 μM). Exhibits 100-fold average selectivity for the tumor cells versus human foreskin fibroblasts. Inhibits EGFR/ErbB2 autophosphorylation and phosphorylation of the downstream modulator AKT.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine
CAS 231277-92-2
Quantity 10 mg
Product Line Tocris
Molecular Formula C29H26ClFN4O4S
Formula Weight 581.06
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.