missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Lapatinib​

Product Code. 17130192
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17130192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130192 Supplier STEMCELL Technologies Supplier No. 73242
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits EGFR and HER2

  • Lapatinib is a selective dual inhibitor of epidermal growth factor receptor (EGFR, ERBB1) and human epidermal growth factor receptor 2 (HER2, ERBB2), each with an IC50 of about 10 nM (Rusnak et al.) It binds reversibly, but with a very slow off-rate, to the ATP binding pocket in an inactive-like conformation, thus preventing autophosphorylation
  • Binding is very potent, with Ki values of 3 and 13 nM to EGFR and HER2 respectively (Wood et al.)
  • Induces apoptosis in cells overexpressing EGFR and/or HER2, and inhibits tumor growth in a mouse xenograft model (Xia et al.)

Specifications

CAS 231277-92-2
Synonym GSK 572016; GW 572016; Tyverb; GW2016
Quantity 10 mg
Molecular Formula C29H26ClFN4O4S
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.