missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib-d7 Sulfoxide Dihydrochloride, TRC

Product Code. 30477516
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477516 50 mg 50mg
30475727 100 mg 100mg
30483567 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477516 Supplier Toronto Research Chemicals Supplier No. TRCL17581250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lapatinib-d7 Sulfoxide Dihydrochloride
Molecular Formula C29H19D7ClFN4O3S . 2(HCl)
Quantity 50 mg
Synonym N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfinyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine-d7 Dihydrochloride
InChI Formula InChI=1S/C29H26ClFN4O3S.2ClH/c1-39(36)12-11-32-16-23-7-10-27(38-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)37-17-19-3-2-4-21(31)13-19;;/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35);2*1H/i1D3,11D2,12D2;;
SMILES FC1=CC(COC(C(Cl)=C2)=CC=C2NC3=NC=NC4=CC=C(C5=CC=C(CNC([2H])([2H])C([2H])([2H])S(C([2H])([2H])[2H])=O)O5)C=C43)=CC=C1.Cl.Cl
IUPAC Name N-(3-chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-((methyl-d3)sulfinyl)ethyl-1,1,2,2-d4)amino)methyl)furan-2-yl)quinazolin-4-amine dihydrochloride
Molecular Weight (g/mol) 645.02
Formula Weight 643.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.