missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib Ditosylate, TRC

Product Code. 30135357
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135357 25 mg 25mg
17989949 5 mg 5mg
18003130 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135357 Supplier Toronto Research Chemicals Supplier No. TRCL17580025MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 388082-77-7
Molecular Formula C29 H26 Cl F N4 O4 S . 2 C7 H8 O3 S
Molecular Weight (g/mol) 925.46
Synonym 4-Quinazolinamine, N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-, 4-methylbenzenesulfonate (1:2), 4-Quinazolinamine, N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-, bis(4-methylbenzenesulfonate) (9CI), GW 572016F, Lapatinib ditosylate, Tykerb
IUPAC Name N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[(2-methylsulfonylethylamino)methyl]furan-2-yl]quinazolin-4-amine;4-methylbenzenesulfonic acid
SMILES Cc1ccc(cc1)S(=O)(=O)O.Cc2ccc(cc2)S(=O)(=O)O.CS(=O)(=O)CCNCc3oc(cc3)c4ccc5ncnc(Nc6ccc(OCc7cccc(F)c7)c(Cl)c6)c5c4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C29H26ClFN4O4S.2C7H8O3S/c1-40(36,37)12-11-32-16-23-7-10-27(39-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)38-17-19-3-2-4-21(31)13-19;2*1-6-2-4-7(5-3-6)11(8,9)10/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35);2*2-5H,1H3,(H,8,9,10)
Formula Weight 924.1736
Chemical Name or Material Lapatinib Ditosylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.