Learn More
Thermo Scientific™ Large White Labels for DYMO Printer
Product Code. 31921088 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Dymo Label Writer 450 printer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
31921088 Dymo Label Writer 450 printer Each
1 options
Product Code. 31921088 Supplier Thermo Scientific™ Supplier No. 869177000
The Thermo Scientific™ NanoDrop™ Dymo LabelWriter 450 Printer is a convenient accessory for NanoDrop instruments. It prints sample data onto self-adhesive labels, which are included with purchase of printer.
The Dymo LabelWriter 550 Turbo label printer with labels for the Thermo Scientific™ NanoDrop™ is a convenient accessory for our microvolume UV-Vis instruments. It quickly and easily prints sample data onto self-adhesive labels, which are included with purchase of printer.
Features include:
  • Generates 2.31 x 4 inch (59mm x 102mm) self-adhesive labels, allowing you to place labels directly in laboratory notebooks
  • Prints results for single samples or batches of samples
  • Direct thermal label printer, no ink or toner required
LabelWriter is a registered trademark of DYMO Corporation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description Large White Labels for DYMO Printer
Dimensions (L x W) 2.3 x 4 in. (59 x 101 cm)
For Use With (Equipment) Dymo Label Writer 450 printer
Packaging 300 labels per roll