missing translation for 'onlineSavingsMsg'
Learn More

Larotaxel, TRC

Product Code. 30132591
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132591 25 mg 25mg
30004411 10 mg 10mg
30003762 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132591 Supplier Toronto Research Chemicals Supplier No. TRCL17600025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156294-36-9
Molecular Formula C45 H53 N O14
Molecular Weight (g/mol) 831.9
Synonym (αR,βS)-β-[[(1,1-Dimethylethoxy)carbonyl]amino]-α-hydroxybenzenepropanoic Acid (1S,2S,4S,7R,8aR,9aS,10aR,12aS,12bR)-7,12a-Bis(acetyloxy)-1-(benzoyloxy)-1,3,4,7,8,9,9a,10,10a,12,12a,12b-dodecahydro-2-hydroxy-5,13,13-trimethyl-8-oxo-2,6-methano-2H-cyclodeca[3,4]cyclopropa[4,5]benz[1,2-b]oxet-4-yl Ester, PNU 100940;
SMILES CC(=O)O[C@H]1C(=O)[C@]23C[C@H]2C[C@H]4OC[C@@]4(OC(=O)C)[C@H]3[C@H](OC(=O)c5ccccc5)[C@]6(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c7ccccc7)C(=C1C6(C)C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C45H53NO14/c1-23-29(57-39(52)33(49)32(26-15-11-9-12-16-26)46-40(53)60-41(4,5)6)21-45(54)37(58-38(51)27-17-13-10-14-18-27)35-43(36(50)34(56-24(2)47)31(23)42(45,7)8)20-28(43)19-30-44(35,22-55-30)59-25(3)48/h9-18,28-30,32-35,37,49,54H,19-22H2,1-8H3,(H,46,53)/t28-,29+,30-,32+,33-,34-,35+,37+,43-,44+,45-/m1/s1
Formula Weight 831.3466
Chemical Name or Material Larotaxel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.