missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Latanoprost

Product Code. 17261262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17261262 100 mg 100mg
17251262 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17261262 Supplier Thermo Scientific Chemicals Supplier No. 466661000
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130209-82-4 | C26H40O5, C26H40O5 | 432.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130209-82-4
Molecular Formula C26H40O5

Specifications

Quantity 100 mg
Solubility Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol and acetonitrile
Formula Weight 432.6
Chemical Name or Material Latanoprost
compliance-icons compliance-icons
Product Identifier
  • Latanoprost
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Latanoprost

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.