missing translation for 'onlineSavingsMsg'
Learn More

Latanoprost Tris(triethylsilyl) Ether, TRC

Product Code. 30473519
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473519 10 mg 10mg
30466338 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473519 Supplier Toronto Research Chemicals Supplier No. TRCL17739010MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477884-78-9
Molecular Formula C44 H82 O5 Si3
Molecular Weight (g/mol) 775.38
Synonym (5Z)-7-[(1R,2R,3R,5S)-2-[(3R)-5-phenyl-3-[(triethylsilyl)oxy]pentyl]-3,5-bis[(triethylsilyl)oxy]cyclopentyl]-5-Heptenoic acid 1-methylethyl ester, Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-bis(triethylsilyloxy)-2-((R)-3-(triethylsilyloxy)-5-phenylpentyl)cyclopentyl]hept-5-enoate, Latanoprost Related Compound D (USP)
IUPAC Name propan-2-yl (Z)-7-[(1R,2R,3R,5S)-2-[(3R)-5-phenyl-3-triethylsilyloxypentyl]-3,5-bis(triethylsilyloxy)cyclopentyl]hept-5-enoate
SMILES CC[Si](CC)(CC)O[C@H](CC[C@@H]1[C@@H](C\C=C/CCCC(=O)OC(C)C)[C@H](C[C@H]1O[Si](CC)(CC)CC)O[Si](CC)(CC)CC)CCc2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C44H82O5Si3/c1-12-50(13-2,14-3)47-39(33-32-38-28-24-23-25-29-38)34-35-41-40(30-26-21-22-27-31-44(45)46-37(10)11)42(48-51(15-4,16-5)17-6)36-43(41)49-52(18-7,19-8)20-9/h21,23-26,28-29,37,39-43H,12-20,22,27,30-36H2,1-11H3/b26-21-/t39-,40+,41+,42-,43+/m0/s1
Formula Weight 774.547
Chemical Name or Material Isopropyl (Z)-7-{(1R,2R,3R,5S)-2-[(R)-5-phenyl-3-(triethylsilyloxy)pentyl]-3,5-bis(triethylsilyloxy)cyclopentyl}hept-5-enoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.