Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Clostridium difficile
Product Code. 15968012 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. Quantity unitSize
15968012 50 Tests 50 tests
1 options
Product Code. 15968012 Supplier Novacyt - Microgen Bioproducts Supplier No. M41CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
  • From a range of available technologies for common pathogens and causatives of bowel disease.
  • Specific tests available for a variety of infection detections.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Especificaciones
Content And Storage 2°C to 8°C
Detectable Analytes C. difficile
Type Latex Agglutination Test
Quantity 50 Tests
Test Time 2 min.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.