Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Staphylococcus aureus
Product Code. 15988012 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
100 tests
50 tests
500 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15988012 100 Tests 50 tests
15957112 500 Tests 500 tests
17122199 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15988012 Supplier Novacyt - Microgen Bioproducts Supplier No. M43CE
Quantity Price
1 88.29 € / 50 tests
10 82.82 € / 50 tests
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Content And Storage 2°C to 8°C
Detectable Analytes S. aureus
Type Latex Agglutination Test
Quantity 100 Tests
Test Time 2 min.