Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Streptococcus A, B, C, D, F and G
Product Code. 15928022 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. Quantity Unit Size
15928022 50 Tests 50 tests
1 options
Product Code. 15928022 Supplier Novacyt - Microgen Bioproducts Supplier No. M47CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
  • Specific tests available for a variety of infection detections.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Content And Storage 2°C to 8°C
Detectable Analytes Streptococci (Group A, B, C, D, F, G)
Type Latex Agglutination Test
Quantity 50 Tests
Test Time 2 min.