Learn More
Fisherbrand™ Latex Dropper Teats
Product Code. 12446180 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 mL
1.5 mL
1.5 to 5 mL (Assorted)
2.5 mL
3.75 mL
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12446180 1.5 to 5 mL (Assorted) Pack of 20
12406180 1 mL Pack of 20
12416180 1.5 mL Pack of 20
12426180 2.5 mL Pack of 20
12436180 3.75 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12446180
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For easy dispensing of liquids.
  • Flexible rubber bulbs create suction and dispensing capacity when used with pipettes
  • Provide intuitive manual pipeting control
  • Available in different capacities
Specifications
Material Rubber
Type Bulb
Volume (Metric) 1.5 to 5 mL (Assorted)
Packaging Type Mixed