missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Latex Finger Cots

Product Code. 11726574 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 1440
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11726574 Clear Pack of 1440
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11726574 Supplier Cole-Parmer™ Supplier No. 8160532

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance Meets ASTM requirement D3772 For Physical Properties
Color Clear
Material Latex
Product Type Finger Cot
Size Small
Thickness (Metric) 76.2 mm
For Use With (Application) For Effective Protection Delicate Applications
Quantity 1440Pack
Length (Metric) 69.8 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.