missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Laurdan

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Laurdan is a polarity sensitive fluorescent probe for imaging lipid rafts in membranes (also known as lipid microdomains). It is suitable for use with generalized polarization (GP) imaging and scanning fluorescence correlation spectroscopy (FCS). Laurdan can be used to image lipids in live and fixed cells as well as in whole tissues with multiphoton microscopy. Laurdan has emission maxima of 440 nm and 490 nm in gel and liquid phase membranes, respectively.
Specifications
Specifications
| Chemical Name or Material | 1-[6-(Dimethylamino)-2-naphthalenyl]-1-dodecanone |
| CAS | 74515-25-6 |
| Quantity | 10 mg |
| Target | Fluorescent Lipids Probes and Cell Membrane Stains |
| Molecular Formula | C24H35NO |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction