missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lauric acid, 99%

Product Code. 10469080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Plastic bottle
Unit Size:
100g
1kg
500g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10469080 100 g Plastic bottle 100g
10495272 5 g Glass bottle 5g
10387340 500 g Plastic bottle 500g
10398103 1 kg Plastic bottle 1kg
10751784 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10469080 Supplier Thermo Scientific Chemicals Supplier No. 167281000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143-07-7 | C12H24O2 | 200.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-07-7
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00002736
InChI Key POULHZVOKOAJMA-UHFFFAOYSA-N
Synonym lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
PubChem CID 3893
ChEBI CHEBI:30805
IUPAC Name dodecanoic acid
SMILES CCCCCCCCCCCC(=O)O

Specifications

Melting Point 44.0°C to 46.0°C
Color White
Boiling Point 225.0°C (100.0 mmHg)
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)10CO2H
Quantity 100 g
Beilstein 02, 359
Merck Index 15, 5439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,ether and alcohol
Formula Weight 200.32
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Lauric acid
Show More Show Less
compliance-icons
Product Identifier
  • Lauric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.