missing translation for 'onlineSavingsMsg'
Learn More

Lauric Acid, TRC

Product Code. 30012013
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012013 25 g 25g
30006082 10 g 10g
30007658 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012013 Supplier Toronto Research Chemicals Supplier No. TRCL17855025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-07-7
Molecular Formula C12 H24 O2
Molecular Weight (g/mol) 200.32
Synonym Dodecanoic acid, Lauric acid (8CI), 1-Dodecanoic acid, 1-Undecanecarboxylic acid, ABL, Aliphat No. 4, ContraZeck, Dodecylic acid, Edenor 12/98-100, Edenor C 12, Edenor C 1298-100, Emery 651, Hystrene 9512, Imex C 1299, Kortacid 1299, Laurostearic acid, Lunac L 70, Lunac L 98, NAA 122, NAA 312, NSC 5026, Neo-Fat 12, Neo-Fat 12-43, Nissan NAA 122, Palmac 98-12, Philacid 1200, Prifac 2920, Prifac 2922, Prifrac 2920, Univol U 314, Vulvic acid, n-Dodecanoic acid
IUPAC Name dodecanoic acid
SMILES CCCCCCCCCCCC(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)
Formula Weight 200.1776
Chemical Name or Material Lauric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.