missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl Chloride, TRC

Product Code. 30475860
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475860 1 L 1 L
30470716 250 mL 250mL
30466892 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475860 Supplier Toronto Research Chemicals Supplier No. TRCL1786301L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.76
Synonym Dodecanoic Acid Chloride, Lauric Acid Chloride, n-Dodecanoyl Chloride
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(=O)Cl

Specifications

Quantity 1 L
InChI Formula InChI=1S/C12H23ClO/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3
Formula Weight 218.1437
Chemical Name or Material Lauroyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.