missing translation for 'onlineSavingsMsg'
Learn More

Lauroylcholine Chloride, TRC

Product Code. 30481803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481803 1 g 1g
30471457 5 g 5g
30472147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481803 Supplier Toronto Research Chemicals Supplier No. TRCL1850001G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25234-60-0
Molecular Formula C17 H36 N O2 . Cl
Molecular Weight (g/mol) 321.93
IUPAC Name 2-dodecanoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCC(=O)OCC[N+](C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H36NO2.ClH/c1-5-6-7-8-9-10-11-12-13-14-17(19)20-16-15-18(2,3)4;/h5-16H2,1-4H3;1H/q+1;/p-1
Formula Weight 321.2435
Chemical Name or Material Lauroylcholine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.