missing translation for 'onlineSavingsMsg'
Learn More

Lauroylsarcosine Sodium Salt, TRC

Product Code. 30481601
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481601 10 g 10g
30477869 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481601 Supplier Toronto Research Chemicals Supplier No. TRCL18500510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15 H28 N O3 . Na
Molecular Weight (g/mol) 293.38
Synonym Glycine, N-methyl-N-(1-oxododecyl)-, sodium salt (1:1), Glycine, N-methyl-N-(1-oxododecyl)-, sodium salt (9CI), Sarcosine, N-lauroyl-, sodium salt (8CI), Compound 105, Crodasinic LS 30, Crodasinic LS 95, Crodasinic LS30NP, Enagicol L 30N, FS 701, Firet L, GM 9011, Gardol, Gardol (antiseptic), Hamposyl L 30, Hamposyl L 95, Lauroyl sarcosine sodium, Lauroylsarcosine sodium salt, Maprosyl 30, Maprosyl 30B, Medialan LD, Medialan LL 99, N-Dodecanoyl-N-methylglycine sodium salt, N-Dodecanoylsarcosine sodium salt, N-Lauroyl-N-methylglycine sodium salt, N-Lauroylsarcosine sodium, N-Lauroylsarcosine sodium salt, Neoscoap SLN 100, Nikkol Sarcosinate LN, Nikkol Sarcosinate LN 3, Nikkol Sarcosinate LN 30, Nikkol Sarcosine Na, Oramix L 30, Perlastan L 30, Protelan LS 9011, SKL, SKL (salt), Sarcosinate LN, Sarcosinate LN 3, Sarcosinate LN 30, Sarkosyl, Sarkosyl NL, Sarkosyl NL 100, Sarkosyl NL 30, Sarkosyl NL 35, Sarkosyl NL 97, Secosyl, Sodium Lauroyl Sarcosinate, Sodium N-lauroylsarcosinate, Sodium N-lauroylsarcosine, Sodium lauroylsarcosine, Soypon SLE, Soypon SLP
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1
Formula Weight 293.1967
Chemical Name or Material Sodium lauroylsarcosinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.