missing translation for 'onlineSavingsMsg'
Learn More

Lauryl chloride, 99%

Product Code. 10056103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10056103 1 L Glass bottle 1 L
10367990 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10056103 Supplier Thermo Scientific Acros Supplier No. 257100010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-52-7 | C12H25Cl | 204.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-52-7
Molecular Formula C12H25Cl
Molecular Weight (g/mol) 204.78
MDL Number MFCD00013688
InChI Key YAYNEUUHHLGGAH-UHFFFAOYSA-N
Synonym lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk
PubChem CID 8192
IUPAC Name 1-chlorododecane
SMILES CCCCCCCCCCCCCl

Specifications

Melting Point -9.3°C
Color Colorless
Density 0.8700g/mL
Boiling Point 260°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11Cl
Refractive Index 1.4420 to 1.444
Quantity 1 L
Specific Gravity 0.87
Solubility Information Solubility in water: 0.0068g/L (20°C). Other solubilities: miscible with most common organic solvents,slightly miscible with alcohol,soluble in ethanol,benzene,ether,miscible with acetone
Formula Weight 204.79
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Lauryl chloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lauryl chloride
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.