missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lauryldimethylamine-N-oxide (LDAO), LC/MS grade

Product Code. 30245091 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30245091 1 g 1g
30245145 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30245091 Supplier Thermo Scientific™ Supplier No. A65503
  • ONLINE EXCLUSIVE PRICE
    81.50 €
    70.91 € / 1g
    Save 10.59 €
    12% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lauryldimethylamine-N-oxide (LDAO) is a zwitterionic surfactant that possesses unique properties for solubilization and stabilization of biomolecules, particularly membrane proteins.

Lauryldimethylamine-N-oxide (LDAO) is a zwitterionic surfactant that possesses unique properties for solubilization and stabilization of biomolecules, particularly membrane proteins. LDAO shows particular utility in native mass spectrometry (MS) to study the intact structure and non-covalent interactions of proteins and protein complexes. Incorporation of LDAO into sample preparation can help maintain the integrity of non-covalent interactions, such as protein-protein or protein-ligand interactions, during the transfer from solution to gas phase.

Features of LDAO include:
• Zwitterionic detergent
• Mildness and biocompatibility
• Enhances the stability of solubilized proteins, including membrane proteins
• High-purity compound with low UV absorptivity
• Native mass spec compatible

Applications
LDAO finds applications in various experimental techniques in addition to native MS, including protein purification, crystallization, and structural biology studies. It is compatible with a wide range of biomolecules and can be employed for both soluble and membrane protein research.

Usage
It is important to optimize the concentration of LDAO to ensure that it provides effective solubilization and stabilization of the biomolecules while avoiding excessive detergent interference in the mass spectrometry analysis. Additionally, the specific conditions and parameters of the native MS experiment, such as the choice of ionization source and mass spectrometer parameters, should be carefully considered and optimized for each specific application.

Properties
CMC: (H2O) ~1–2 mM (0.023 %)
Aggregation number: (H2O) ~76
Purity: ≥99% by HPLC analysis (all amine oxides) with <2% other DAOs
pH: 6–8.5 (1% solution in water)
Solubility: ≥30% (in water at 20°C)
Fluorescence: <10% (0.1 solution in water at 345 nm)
Absorbance of a 1% solution in water: 260 nm: <0.08, 280 nm: <0.06, 340 nm: <0.02

TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Lauryldimethylamine-N-oxide
CAS 1643-20-5
Molecular Formula C14H31NO
Form Powder
For Use With (Application) LC-MS
Quantity 1 g
Molecular Weight (g/mol) 229.4
Purity ≥99%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.