missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals LB Broth (Miller's modification)

Product Code. 11372599
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging Type:
Glass Bottle
Plastic Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Unit Size
11372599 500 g Plastic Bottle 500g
11362599 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372599 Supplier Thermo Scientific Chemicals Supplier No. H26676.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

(+)-Dehydroisoandrosterone, CAS # 53-43-0, also known as androstenolone, is an endogenous steroid hormone precursor. It is an intermediate in the conversion of cholesterol to androgens and estrogens.

For the growth of E. coli and other enteric bacteria. LB Luria contains less salt than LB Miller and LB Lennox. LB Luria is used for cultivating salt-sensitive strains of E. coli and enteric bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • (+)-Dehydroisoandrosterone is an endogenous steroid hormone precursor that acts as an intermediate in the conversion of cholesterol to androgens and estrogens

Applications

  • It is an endogenous and naturally occurring steroid hormone produced largely by the adrenal glands, but also by the testes and ovaries
  • It has been described that (+)-Dehydroisoandrosterone could have a role in the immune and stress response
  • (+)-Dehydroisoandrosterone can be converted to testosterone, androstenedione, estradiol, and estrone
  • (+)-Dehydroisoandrosterone is derived from cholesterol through the action of two cytochrome P450 enzymes
  • It can induce both direct and indirect in vivo biological effects, by modulating GABA- α receptor and N-Methyl-D-aspartic acid (NMDA) receptor complexes.
  • It can also increase pancreatic β cell insulin secretion and anti-glucocorticoid activity
  • It is useful to detect excess adrenal activity since it is produced nearly exclusively by the adrenal glands
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Packaging Type Plastic Bottle
Quantity 500 g
Sensitivity Hygroscopic
How should I prepare 1 L of Miller LB Broth media (Cat. Nos. 611875000 and H26676.xx)?

To prepare 1 L of Miller LB media:

  1. Add 25 g of powder to 1 L distilled water
  2. Mix by vortexing
  3. Sterilize by autoclaving for 25 minutes at 121 degrees C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.