missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ LC PEEKsil™ Tubing

Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.

Brand:  Trajan Scientific and Medical™ 0624302

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288763

  • 104.00 € / Pack of 5
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Specifications

Specifications

Tubing
1/16 in.
1.6 mm
Polymer-sheathed Fused Silica
5/Pk.
0.1 mm
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers