Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.
44.10 € - 117.00 €
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | |||
15258763
|
Trajan Scientific and Medical™
0624293 |
1.6 mm | 0.075 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15258783
|
Trajan Scientific and Medical™
0624365 |
1.6 mm | 0.53 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15268733
|
Trajan Scientific and Medical™
0624217 |
1.6 mm | 0.3 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15268743
|
Trajan Scientific and Medical™
0624232 |
1.6 mm | 0.15 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15268753
|
Trajan Scientific and Medical™
0624254 |
1.6 mm | 0.05 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15268763
|
Trajan Scientific and Medical™
0624300 |
1.58 mm | 0.1 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15298763
|
Trajan Scientific and Medical™
0624303 |
1.58 mm | 0.1 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15248743
|
Trajan Scientific and Medical™
0624233 |
1.6 mm | 0.15 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15298753
|
Trajan Scientific and Medical™
0624263 |
0.8 mm | 0.05 mm | 150 mm |
44.10 €
Pack of 2 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
11991606
|
Trajan Scientific and Medical™
0624218 |
1.6 mm | 0.3 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||
15208773
|
Trajan Scientific and Medical™
0624304 |
1.6 mm | 0.1 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15258733
|
Trajan Scientific and Medical™
0624216 |
1.6 mm | 0.3 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15268783
|
Trajan Scientific and Medical™
0624367 |
1.6 mm | 0.53 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15238763
|
Trajan Scientific and Medical™
0624291 |
1.6 mm | 0.075 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15278733
|
Trajan Scientific and Medical™
0624225 |
1.6 mm | 0.025 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15298733
|
Trajan Scientific and Medical™
0624227 |
1.6 mm | 0.025 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
15208733
|
Trajan Scientific and Medical™
0624204 |
1.6 mm | 0.2 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15208743
|
Trajan Scientific and Medical™
0624228 |
1.6 mm | 0.025 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15218733
|
Trajan Scientific and Medical™
0624205 |
1.6 mm | 0.2 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15218743
|
Trajan Scientific and Medical™
0624229 |
1.6 mm | 0.025 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
15228743
|
Trajan Scientific and Medical™
0624230 |
1.6 mm | 0.15 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15228763
|
Trajan Scientific and Medical™
0624290 |
1.6 mm | 0.075 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15238733
|
Trajan Scientific and Medical™
0624214 |
1.6 mm | 0.3 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15238743
|
Trajan Scientific and Medical™
0624231 |
1.6 mm | 0.15 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15238783
|
Trajan Scientific and Medical™
0624354 |
1.6 mm | 0.175 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15248733
|
Trajan Scientific and Medical™
0624215 |
1.6 mm | 0.3 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15248763
|
Trajan Scientific and Medical™
0624292 |
1.6 mm | 0.075 mm | 150 mm |
111.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15248783
|
Trajan Scientific and Medical™
0624366 |
1.6 mm | 0.53 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15278723
|
Trajan Scientific and Medical™
0624201 |
1.6 mm | 0.2 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15278763
|
Trajan Scientific and Medical™
0624301 |
1.6 mm | 0.1 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15278783
|
Trajan Scientific and Medical™
0624370 |
1.6 mm | 0.53 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15288723
|
Trajan Scientific and Medical™
0624202 |
1.6 mm | 0.2 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15288733
|
Trajan Scientific and Medical™
0624226 |
1.6 mm | 0.025 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15288763
|
Trajan Scientific and Medical™
0624302 |
1.6 mm | 0.1 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15298723
|
Trajan Scientific and Medical™
0624203 |
1.6 mm | 0.2 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
15258743
|
Trajan Scientific and Medical™
0624234 |
1.6 mm | 0.15 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Tubing | |
Polymer-sheathed Fused Silica |