missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Shop All Macherey-Nagel Products
Click to view available options
Chemical Name or Material:
2'7'-Dichlorofluorescein
4-(Dimethylamino)-benzaldehyde
Aniline phthalate
Bromocresol green
Caffeine detection reagent
Dragendorff-Munier
Iron(III) Chloride
Molydatophosphoric acid
Ninhydrin
Potassium Ferricyanide
Rhodamin B
Rubeanic acid
Unit Size:
1 set
100mL
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
Specifications
| Chemical Name or Material | Caffeine detection reagent |
| CAS | 7705-08-0 |
| Color | Slightly yellow |
| Detection Method | Spraying or Dipping |
| Formulation | Iodine / Iron(III) Chloride / L(+)-Tartaric Acid in Water / Acetone |
| For Use With (Application) | Caffeine-Detection |
| For Use With (Equipment) | TLC plates |
| Matrix | Water-Acetone |
| Odor | Odorless |
| Packaging Type | Amber bottle with screw cap |
| Show More |
Safety and Handling
Product Identifier
- 100ML Spray reagent for caffeine L
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : acetone, iodine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction