missing translation for 'onlineSavingsMsg'
Learn More

Hach™ LCK Cuvette Tests

Product Code. 10182404
Change view
Click to view available options
Analytes:
Acid Capacity (KS4.3/CaCO3)
Boron
Dissolved Sulphide
Molybdenum
Nitrite
Nonionic Surfactants
Total Organic Carbon
Unit Size:
25 tests
Each
Pack of 25
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analytes Unit Size
10182404 Dissolved Sulphide Pack of 25
10656090 Molybdenum Each
10122744 Acid Capacity (KS4.3/CaCO3) Each
10156112 Nonionic Surfactants Each
11578149 Total Organic Carbon 25 tests
10137309 Boron Each
10749873 Nitrite Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10182404 Supplier Hach™ Supplier No. LCK653
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Insert, read and finish. Hach™ LCK Cuvette Tests are pre-dosed reagents packaged in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation, while using a minimum amount of reagent/chemicals for an environmentally friendly solution.

Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.

  • Pre-dosed, ready-to-use reagents with no rinsing steps needed
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Order Info

dimProductType

TRUSTED_SUSTAINABILITY

Specifications

Analytes Dissolved Sulphide
Certifications/Compliance ISO 10530-1991, DIN 38405-D26
Color Light blue
Format Pre-dosed Tubes
For Use With (Application) Water quality testing
For Use With (Equipment) Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions
Includes Fully equipped box, reagent set
No. of Tests 25
Range 0.1 to 2.0mg/L S2-
Storage Requirements 15°C to 25°C
Test Method Dimethyl-p-phenylenediamine
Type Sulphide Cuvette Test
Show More Show Less
compliance-icons
Product Identifier
  • STABLCAL Turbidity Standards Calibration Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid, N,N-Dimethyl-1,4-phenylenediammonium dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.