missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LDN 209929 dihydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
LDN 209929 dihydrochloride is a potent and selective haspin kinase inhibitor (IC50 = 55 nM). Displays 180-fold selectivity over DYRK2.
Specifications
Specifications
| Purity | >98% |
| CAS | 1784281-97-5 |
| Quantity | 50 mg |
| Product Type | LDN 209929 dihydrochloride |
| Molecular Weight (g/mol) | 405.77 |
| Inhibitors | LDN 209929 dihydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction