missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LDN 209929 dihydrochloride

Product Code. p-5545860
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18784729

Brand: Tocris Bioscience™ 4828/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective haspin inhibitor

LDN 209929 dihydrochloride is a potent and selective haspin kinase inhibitor (IC50 = 55 nM). Displays 180-fold selectivity over DYRK2.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
CAS 1784281-97-5
Quantity 10 mg
Product Type LDN 209929 dihydrochloride
Molecular Weight (g/mol) 405.77
Inhibitors LDN 209929 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.