Learn More
Tocris Bioscience™ LDN 209929 dihydrochloride
Potent and selective haspin inhibitor
Brand:  Tocris Bioscience™ 4828/10
Encompass
Product Code. 15726585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
LDN 209929 dihydrochloride is a potent and selective haspin kinase inhibitor (IC50 = 55 nM). Displays 180-fold selectivity over DYRK2.
Specifications
Specifications
>98%
1784281-97-5