missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE

Product Code. 15778877
Change view
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Closure Color:
Blue
Green
Orange
Red
Yellow
Unit Size:
Each
Pack of 10
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Label Closure Color Unit Size
15778877 Isopropanol Yellow Pack of 10
10667691 Acetone Red Each
15708887 Acetone Red Pack of 10
15768877 Ethanol Green Pack of 10
15788877 Methanol Orange Pack of 10
17898097 Methanol Orange Pack of 100
15798877 Distilled Water Blue Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15778877 Supplier Kautex™ Supplier No. 2000770022
  • ONLINE EXCLUSIVE PRICE
    78.89 €
    71.63 € / Pack of 10
    Save 7.26 €
    9% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to give an accurate fine controlled stream of liquid. Kautex™ 500mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.

  • Imprinted with hazard symbols and substances of content in German, English and French language
  • Writing space
  • Abrasion-resistant printing
  • Closure color matches color of ink for easy identification of media
  • Leak-proof
  • Completely emptied by sliding delivery tube, even when inverted.
Suitable for the declared liquids. Not suitable for food.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Natural
Diameter (Metric) Inner Neck 39.2 mm
Material LDPE
No. per Pack 10
Quantity 100 Pk.
Closure Color Yellow
Mouth Wide
Height (Metric) 154 mm
Chemical Label Isopropanol
Diameter (Metric) 77 mm
Closure Type Screw Cap with Delivery Nozzle
Shape Round
Product Type Wash Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.