missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Natural |
| Diameter (Metric) Inner Neck | 39.2 mm |
| Material | LDPE |
| No. per Pack | 10 |
| Quantity | 100 Pk. |
| Closure Color | Blue |
| Mouth | Wide |
| Height (Metric) | 154 mm |
| Chemical Label | Distilled Water |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?