missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE

Designed to give an accurate fine controlled stream of liquid. Kautex™ 500mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.

Brand:  Kautex™ 2000770023

Additional Details : Weight : 5.30000kg

 View more versions of this product

Product Code. 17898097

  • 646.25 € / Pack of 100
Estimated Shipment: 22-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Imprinted with hazard symbols and substances of content in German, English and French language
  • Writing space
  • Abrasion-resistant printing
  • Closure color matches color of ink for easy identification of media
  • Leak-proof
  • Completely emptied by sliding delivery tube, even when inverted.
Suitable for the declared liquids. Not suitable for food.
Specifications

Specifications

Wash Bottle
77 mm
Methanol
Natural
LDPE
100 Pk.
Wide
154 mm
Screw Cap with Delivery Nozzle
500 mL
39.2 mm
10
Orange
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers