missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ LDPE Lids

Product Code. p-8000502
Click to view available options
For Use With (Equipment):
For 10 L Bucket
For 5 L Bucket
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10007890

Brand: Vitlab™ 96593

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured with LDPE for tight fit and transparency. VITLab™ LDPE Lids are for use with VITlab™ HDPE buckets.

  • Tightly closing
  • Order buckets separately 

Specifications

Product Type Lid
Material LDPE
Color Transparent
For Use With (Equipment) For 10 L Bucket
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.