missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate Basic, TRC

Product Code. 30483011
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30483011 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30483011

Brand: Toronto Research Chemicals TRCL17878550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead Acetate Basic
CAS 51404-69-4
Molecular Formula C4H8O6Pb2
Quantity 50 g
Synonym Acetic acid, lead salt, basic, Lead acetate basic, Lead(II) acetate basic
InChI Formula InChI=1S/2C2H4O2.2H2O.2Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);2*1H2;;/q;;;;2*+2/p-4
SMILES CC(O[Pb]OC(C)=O)=O.O[Pb]O
Molecular Weight (g/mol) 566.5
Formula Weight 567.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.