missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate Cotton, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11424699 Alle ansehen Reagecon Produkte
Ansicht ändern
Click to view available options
Quantity:
10 g
Packaging:
Bottle, tamper-evident
Packungsgröße:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Packungsgröße
11424699 10 g Bottle, tamper-evident 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11424699 Lieferant Reagecon Lieferanten-Nr. 1048101
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Lead Acetate Cotton, EP Grade, Reagecon™ is a ready-to-use, dried cotton that has absorbed a 10:1 lead acetate-dilute acetic acid solution for use in arsenic limit tests.

Lead acetate cotton is designed for use in arsenic limit tests. Absorbent cotton is immersed in a mixture of 1 part dilute acetic acid + 10 parts lead acetate solution, placed on layers of filter paper to drain, and allowed to air dry.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Spezifikation

CAS 6080-56-4, 64-19-7
Color White
Odor Odorless
Packaging Bottle, tamper-evident
Molecular Formula C4H12O7Pb
MDL Number MFCD00150023
Quantity 10 g
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
IUPAC Name lead(2+);diacetate;trihydrate
Molecular Weight (g/mol) 379.30
PubChem CID 22456
ChEBI CHEBI:33112
Formula Weight 379.3g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Concentration or Composition (by Analyte or Components) 1 dilute acetic acid:10 lead acetate solution
Chemical Name or Material Lead di-acetate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • LEAD ACETATE COTTON EUROPEAN PHARMACOPOEIA
Signalwort
  • Danger
Gefahrenkategorie
  • Reproductive toxicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 2
Gefahrenhinweis
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH201-Contains lead. Should not be used on surfaces liable to be chewed or sucked by children.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Ergänzende Informationen
  • MIXTURE LIST-Contain: lead di(acetate), acetic acid
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.