missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate Trihydrate, TRC

Product Code. 30467035
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467035 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467035 Supplier Toronto Research Chemicals Supplier No. TRCL3200605G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead Acetate Trihydrate
CAS 6080-56-4
Molecular Formula C4H6O4Pb . 3H2O
Quantity 5 g
Synonym Lead(II) Acetate Trihydrate
InChI Formula InChI=1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2
SMILES O.O.O.[Pb+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name lead(2+);diacetate;trihydrate
Molecular Weight (g/mol) 379.33
Formula Weight 380.0349
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.