missing translation for 'onlineSavingsMsg'
Learn More

Lead citrate, trihydrate, 96%, Thermo Scientific™

Product Code. 10157420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157420 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157420 Supplier Thermo Scientific Acros Supplier No. 413061000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6107-83-1
Molecular Formula C12H16O17Pb3
Molecular Weight (g/mol) 1053.843
MDL Number MFCD00150456
InChI Key GMPMGSCJCDAUMP-UHFFFAOYSA-H
Synonym lead ii citrate trihydrate, acmc-20akng, citric acid lead ii salt trihydrate, tris ? 2-lead 2+ dicitrate trihydrate, 2-hydroxypropane-1,2,3-tricarboxylate; lead 2+ ; trihydrate, 1,2,3-propanetricarboxylicacid, 2-hydroxy-, lead 2+ salt, hydrate 2:3:3
PubChem CID 2735068
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;lead(2+);trihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Pb+2].[Pb+2].[Pb+2]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (HOC(COO-)(CH2COO-)2)2Pb3·3H2O
Quantity 100 g
Formula Weight 1053.85
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Lead citrate, trihydrate, 96%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead citrate, trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.