missing translation for 'onlineSavingsMsg'
Learn More

Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)

Product Code. 15435905
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435905 1 Pc. Each
15445905 5 Pc. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435905 Supplier Thermo Scientific Alfa Aesar Supplier No. 000708.KF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.1630g/30 x 30cm | CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Appearance Gray foil
Quantity 1 Pc.
Odor Odorless
Assay Percent Range >99% (Metals basis)
Form Foil
Chemical Name or Material Lead
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead foil
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.