Learn More
STERIS™ Lead-free Autoclave Indicator Tape
Description
- Self-adhering tape with a chemical indicator printed on one side
- The easily recognizable stripes change color when exposed to the specific sterilization, verifying exposure to the process without the need to open packs or check records
- Color change brown/black
Specifications
Specifications
| Color | Brown, Black |
| Dimensions (LxW) | 50 m x 25 mm |