missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) 2,4-pentanedionate

Product Code. 11433840
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433840 50 g 50g
11423840 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433840 Supplier Thermo Scientific Alfa Aesar Supplier No. 089858.18
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15282-88-9 | C10H14O4Pb | 405.418 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15282-88-9
Molecular Formula C10H14O4Pb
Molecular Weight (g/mol) 405.418
MDL Number MFCD00013499
InChI Key UNNUWSQNTAFLDC-SYWGCQIGSA-L
Synonym lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
PubChem CID 53393517
IUPAC Name bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
SMILES CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C

Specifications

Melting Point 141°C to 144°C
Odor Odorless
Quantity 50 g
UN Number UN3467
Sensitivity Hygroscopic
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 405.41
Physical Form Crystalline
Chemical Name or Material Lead (II) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.