missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, 99+%, for analysis

Product Code. p-44101 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10133930

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.0 to 103% (Complexometry)
Linear Formula (CH3CO2)2Pb·3H2O
Merck Index 15, 5452
Solubility Information Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
IUPAC Name λ²-lead(2+) diacetate trihydrate
Formula Weight 379.33
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Lead(II) acetate trihydrate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.