missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Acetate Trihydrate, ACS Reagent, 99.0 to 103.0%, Honeywell Fluka™

Product Code. 15602010 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
25g
500g
Product Code. Quantity Packaging unitSize
15602010 25 g Poly Bottle 25g
15612010 500 g Poly Bottle 500g
15692000 2.5 kg Poly Bottle 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 15602010

Brand: Honeywell-Fluka 21590225G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Boiling Point 204°C
Melting Point 75°C
Color Colorless
Physical Form Solid
Quantity 25 g
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number UN1616
Beilstein 3730298
IUPAC Name λ²-lead(2+) diacetate trihydrate
Formula Weight 379.33g/mol
Percent Purity 99.0 to 103.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.550g/cu.cm
Chemical Name or Material Lead(Ii) Acetate Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.