missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, Puriss., ≥99.5%, Honeywell Fluka™

Product Code. 15651050 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
50 kg
Unit Size:
1kg
25kg
50kg
5kg
This item is not returnable. View return policy

Product Code. 15651050

Brand: Honeywell-Fluka 115041KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 1 kg
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number 1616
Beilstein 3730298
IUPAC Name lead(2+);diacetate;trihydrate
Formula Weight 379.33g/mol
Percent Purity ≥99.5%
Grade puriss.
Chemical Name or Material Lead(Ii) Acetate Trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.