missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) bromide, 98+%, extra pure

Product Code. 10761091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761091 100 g Glass bottle 100g
10092460 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10761091 Supplier Thermo Scientific Acros Supplier No. 198851000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10031-22-8 | Br2Pb | 367.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
SMILES [Br-].[Br-].[Pb++]

Specifications

Boiling Point 916.0°C
Melting Point 373.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Loss on Drying 0.1% max.
Packaging Glass bottle
Specific Gravity 6.66
Assay Percent Range 98% min. (Argentometry)
Density 6.6600g/mL
Linear Formula PbBr2
Fieser 15,182
Merck Index 15, 5458
Solubility Information Solubility in water: 5g/L (20 c). Other solubilities: insoluble in alcohol
IUPAC Name λ²-lead(2+) dibromide
Formula Weight 367.01
Percent Purity ≥98%
Grade Extra Pure
Chemical Name or Material Lead(II) bromide, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Lead bromide (PbBr2)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.