missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Bromide, Pure, Fisher Chemical™

Product Code. 10686452 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10686452 250 g 250g
10686262 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686452 Supplier Fisher Chemical Supplier No. L/1100/50
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
IUPAC Name λ²-lead(2+) dibromide
SMILES [Br-].[Br-].[Pb++]

Specifications

Boiling Point 916°C
Melting Point 373°C
Color White
Physical Form Powder/Solid
Quantity 250 g
Formula Weight 367.01g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.